4-(Dimetyyliamino)bentsoehappo ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

D0724-25G D0724-500G
TCIAD0724-25GEA 23.1 EUR
TCIAD0724-25G TCIAD0724-500G
4-(Dimetyyliamino)bentsoehappo ≥98.0% (by HPLC, titration analysis)
4-(Dimetyyliamino)bentsoehappo
Kaava: C₉H₁₁NO₂
Kiehumispiste: 297 °C (1013 hPa)
Sulamispiste: 243°C
Tiheys: 1,29 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002537
CAS-numero: 619-84-1
EINECS: 210-615-8
Merck Index: 14,03232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=950baf9fdaf4a56eaa95b8986cb8a4bb05d49a13&vid=819cd097cc5f54631ddec73819b8b33a4f2c8e2e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR