N,N-Diethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

D0482-25G D0482-500G D0482-100G
TCIAD0482-25GEA 24.2 EUR
TCIAD0482-25G TCIAD0482-500G TCIAD0482-100G
N,N-Diethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
N,N-Diethylcarbamoyl chloride
Kaava: C₅H₁₀ClNO
Leimahduspiste: 76°C
MDL numero: MFCD00000636
CAS-numero: 88-10-8
EINECS: 201-798-5
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b188062313bf1213309ba13391c5158f1805c97c&vid=b188062313bf1213309ba13391c5158f1805c97c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR