N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

D0469-1G D0469-25G D0469-5G
TCIAD0469-1GEA 17.6 EUR
TCIAD0469-1G TCIAD0469-25G TCIAD0469-5G
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥98.0% (by titrimetric analysis)
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
Kaava: C₁₆H₂₄N₂O₄
MDL numero: MFCD00060156
CAS-numero: 29473-53-8

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR