3a,4,7,7a-Tetrahydro-4,7-metaani-indeeni ≥97.0% (by GC) stabiloitu precursor to cyclopentadiene

Toimittaja: TCI
Vaara


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

D0443-25ML D0443-500ML
TCIAD0443-25MLEA 17.6 EUR
TCIAD0443-25ML TCIAD0443-500ML
3a,4,7,7a-Tetrahydro-4,7-metaani-indeeni ≥97.0% (by GC) stabiloitu precursor to cyclopentadiene
3a,4,7,7a-Tetrahydro-4,7-metaani-indeeni
Kaava: C₁₀H₁₂
Kiehumispiste: 170 °C (1013 hPa)
Sulamispiste: 12…34 °C
Tiheys: 0,982 g/cm³ (20 °C)
Leimahduspiste: 44°C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00078246
CAS-numero: 77-73-6
EINECS: 201-052-9
UN: 2048
ADR: 3,III
Merck Index: 13,02767
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75b9a153d250ae1b6cfce6ba632ba1bc109959eb&vid=42a1d5290aaa7b2464f1bcdfb64319072bacc302&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR