1,3-dibromipropaani ≥98.0% (by GC)

Toimittaja: TCI
Vaara

D0202-25G D0202-500G D0202-100G
TCIAD0202-25GEA 17.6 EUR
TCIAD0202-25G TCIAD0202-500G TCIAD0202-100G
1,3-dibromipropaani ≥98.0% (by GC)
1,3-dibromipropaani
Kaava: BrH₂CCH₂CH₂Br
Kiehumispiste: 167°C
Sulamispiste: –34 °C
Tiheys: 1,99 g/cm³ (20 °C)
Leimahduspiste: 56°C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000255
CAS-numero: 109-64-8
EINECS: 203-690-3
UN: 1993
ADR: 3,III
Merck Index: 14,09713
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef45c0b88630ae5d1263c3e489c22c0ab72fd70e&vid=71cf51d218ea747f6c9bde1c74d02053a6031021&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR