N,N-Dibenzyl-2-aminoethanol ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

D0153-25G D0153-500G
TCIAD0153-25GEA 55 EUR
TCIAD0153-25G TCIAD0153-500G
N,N-Dibenzyl-2-aminoethanol ≥98.0% (by titrimetric analysis)
N,N-Dibenzyl-2-aminoethanol
Kaava: C₁₆H₁₉NO
Kiehumispiste: 206°C
MDL numero: MFCD00020574
CAS-numero: 101-06-4
EINECS: 202-911-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea50ac2e434fd8d066329db89edaf6c06ec27aaf&vid=ea50ac2e434fd8d066329db89edaf6c06ec27aaf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR