(Cyclohexyl-CAAC)Rh(COD)Cl ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

C3592-1G C3592-100MG
TCIAC3592-1GEA 1180 EUR
TCIAC3592-1G TCIAC3592-100MG
(Cyclohexyl-CAAC)Rh(COD)Cl ≥98.0% (by titrimetric analysis)
(Cyclohexyl-CAAC)Rh(COD)Cl
Kaava: C₃₁H₄₇ClNRh
Storage Temperature: Jääkaappi
CAS-numero: 1801869-83-9

Order Now

Testitulosten erittely

Appearance White to Yellow to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR