Schisandrin A ≥98.0% (by GC)

Toimittaja: TCI

C3501-100MG C3501-25MG
TCIAC3501-100MGEA 460 EUR
TCIAC3501-100MG TCIAC3501-25MG
Schisandrin A ≥98.0% (by GC)
Schisandrin A
Kaava: C₂₄H₃₂O₆
Storage Temperature: Jääkaappi
MDL numero: MFCD09026934
CAS-numero: 61281-38-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR