3-(Chlorodimethylsilyl)propyl methacrylate ≥90.0% (by GC) stabiloitu

Toimittaja: TCI
Vaara


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

C3435-5G
TCIAC3435-5GEA 213 EUR
TCIAC3435-5G
3-(Chlorodimethylsilyl)propyl methacrylate ≥90.0% (by GC) stabiloitu
3-(Chlorodimethylsilyl)propyl methacrylate
Kaava: C₉H₁₇ClO₂Si
Kiehumispiste: 82°C
Leimahduspiste: 85°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00053899
CAS-numero: 24636-31-5
EINECS: 246-377-7
UN: 2987
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR