Cesium Pivalate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

C3230-1G C3230-5G
TCIAC3230-1GEA 46.2 EUR
TCIAC3230-1G TCIAC3230-5G
Cesium Pivalate ≥97.0% (by titrimetric analysis)
Cesium Pivalate
Kaava: C₅H₉CsO₂
Sulamispiste: 348°C
MDL numero: MFCD10567040
CAS-numero: 20442-70-0

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR