Carteolol hydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

Synonyms: 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone hydrochloride

C3015-50MG C3015-250MG
TCIAC3015-50MGEA 89.1 EUR
TCIAC3015-50MG TCIAC3015-250MG
Carteolol hydrochloride ≥98.0% (by HPLC, titration analysis)
Carteolol hydrochloride
Kaava: C₁₆H₂₄N₂O₃·HCl
Sulamispiste: 278°C
MDL numero: MFCD00941499
CAS-numero: 51781-21-6
EINECS: 257-415-7
Merck Index: 14,01866

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR