Chlorobis(ethylene)rhodium(I) dimer ≥95.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

C2461-200MG
TCIAC2461-200MGEA 388 EUR
TCIAC2461-200MG
Chlorobis(ethylene)rhodium(I) dimer ≥95.0% (by titrimetric analysis)
Chlorobis(ethylene)rhodium(I) dimer
Kaava: C₈H₁₆Cl₂Rh₂
Storage Temperature: Pakastin
MDL numero: MFCD00013206
CAS-numero: 12081-16-2
EINECS: 235-145-0

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Argentmetric Titration) min. 95.0 %
Rhodium 50.30 to 55.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR