Carbon nanotube multi-walled, Length: 5-15 µm, Ø 20-40 nm

Toimittaja: TCI
Varoitus

C2154-5G C2154-1G
TCIAC2154-5GEA 226 EUR
TCIAC2154-5G TCIAC2154-1G
Carbon nanotube multi-walled, Length: 5-15 µm, Ø 20-40 nm
Carbon nanotube
Kaava: C
MDL numero: MFCD00133992
CAS-numero: 308068-56-6

Order Now

Testitulosten erittely

Appearance Black powder to crystal
Purity (TPO Method) min. 95 %
Diameter 20 to 40 nm
Length 5 to 15 micro_m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR