4'-Chloropivaloanilide ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

C2099-25G C2099-250G
TCIAC2099-25GEA 56.1 EUR
TCIAC2099-25G TCIAC2099-250G
4'-Chloropivaloanilide ≥98.0% (by GC)
4'-Chloropivaloanilide
Kaava: C₁₁H₁₄ClNO
MDL numero: MFCD01571825
CAS-numero: 65854-91-3

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR