4-(Chloromethyl)styrene ≥90.0% (by GC) stabilised

Toimittaja: TCI
Vaara

Synonyms: 1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride

Stabilization: Stabilised with TBC + ONP + 2-nitro-p-cresol

C1792-25G C1792-250G
TCIAC1792-25GEA 42.9 EUR
TCIAC1792-25G TCIAC1792-250G
4-(Chloromethyl)styrene ≥90.0% (by GC) stabilised
4-(Chloromethyl)styrene
Kaava: C₉H₉Cl
Kiehumispiste: 229°C
Leimahduspiste: 103°C
Storage Temperature: Pakastin
MDL numero: MFCD00051362
CAS-numero: 1592-20-7
EINECS: 216-471-2
UN: 3265
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR