(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

C1450-25G C1450-5G
TCIAC1450-25GEA 88 EUR
TCIAC1450-25G TCIAC1450-5G
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate ≥98.0% (by titrimetric analysis)
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate
Kaava: C₆H₁₄N₂
MDL numero: MFCD00191980
CAS-numero: 67333-70-4

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -11.0 to -16.0 deg (C=2.5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR