Diphenylphosphinic chloride ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

C1415-25G C1415-5G
TCIAC1415-25GEA 73.7 EUR
TCIAC1415-25G TCIAC1415-5G
Diphenylphosphinic chloride ≥98.0% (by GC, titration analysis)
Diphenylphosphinic chloride
Kaava: C₁₂H₁₀ClOP
Kiehumispiste: 222°C
Leimahduspiste: 26°C
MDL numero: MFCD00002077
CAS-numero: 1499-21-4
EINECS: 216-107-2
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a34d2dce7aa967d9552127e6e6455842e64e025d&vid=a34d2dce7aa967d9552127e6e6455842e64e025d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR