Carbonylhydridotris(triphenylphosphine)rhodium(I)

Toimittaja: TCI

C1383-1G C1383-5G
TCIAC1383-1GEA 450 EUR
TCIAC1383-1G TCIAC1383-5G
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Kaava: C₅₅H₄₆OP₃Rh
Sulamispiste: 122°C
MDL numero: MFCD00151644
CAS-numero: 17185-29-4
EINECS: 241-230-3
Merck Index: 14,09758

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow powder to crystal
Elemental analysis (Carbon) 70.50 to 72.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR