ω-Bromo-4-cyanoacetophenone ≥96.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

C1348-25G C1348-5G
TCIAC1348-25GEA 95.7 EUR
TCIAC1348-25G TCIAC1348-5G
ω-Bromo-4-cyanoacetophenone ≥96.0% (by GC, titration analysis)
ω-Bromo-4-cyanoacetophenone
Kaava: C₉H₆BrNO
Kiehumispiste: 314 °C (1013 hPa)
Sulamispiste: 92…94 °C
MDL numero: MFCD00052931
CAS-numero: 20099-89-2
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08195a7c89798206ee29e7c4a57be9485a00d251&vid=08195a7c89798206ee29e7c4a57be9485a00d251&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %
Melting point 92.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR