4-(4-Chlorophenyl)piperidin-4-ol ≥97.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

C1291-25G C1291-5G
TCIAC1291-25GEA 246 EUR
TCIAC1291-25G TCIAC1291-5G
4-(4-Chlorophenyl)piperidin-4-ol ≥97.0% (by HPLC, titration analysis)
4-(4-Chlorophenyl)piperidin-4-ol
Kaava: C₁₁H₁₄ClNO
MDL numero: MFCD00006001
CAS-numero: 39512-49-7
EINECS: 254-479-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee0285a9c74faa58eafc380511146276399875c5&vid=ee0285a9c74faa58eafc380511146276399875c5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 138.0 to 142.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR