Sodium chlorodifluoroacetate ≥99.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

C0991-25G C0991-500G C0991-100G
TCIAC0991-25GEA 25.3 EUR
TCIAC0991-25G TCIAC0991-500G TCIAC0991-100G
Sodium chlorodifluoroacetate ≥99.0% (by titrimetric analysis)
Sodium chlorodifluoroacetate
Kaava: C₂ClF₂NaO₂
Sulamispiste: 193…195 °C
MDL numero: MFCD00064771
CAS-numero: 1895-39-2
EINECS: 217-586-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e78d7cba15ce3150ee64c57ed606f44072fdcdc&vid=1e78d7cba15ce3150ee64c57ed606f44072fdcdc&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR