Barium-2-cyanoethylphosphate hydrate ≥98.0% (by titrimetric analysis) phosphorylating agent

Toimittaja: TCI
Vaara

Synonyms: 2-Cyanoethylphosporic Acid Barium Salt

C0978-25G C0978-5G
TCIAC0978-25GEA 150 EUR
TCIAC0978-25G TCIAC0978-5G
Barium-2-cyanoethylphosphate hydrate ≥98.0% (by titrimetric analysis) phosphorylating agent
Barium-2-cyanoethylphosphate hydrate
Kaava: C₃H₆NO₄P
MDL numero: MFCD00012588
CAS-numero: 207121-42-4
UN: 1564
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 5.0 to 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR