L(-)-Camphorsulphonic acid ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

C0972-25G C0972-500G C0972-100G
TCIAC0972-25GEA 22 EUR
TCIAC0972-25G TCIAC0972-500G TCIAC0972-100G
L(-)-Camphorsulphonic acid ≥98.0% (by titrimetric analysis)
L(-)-Camphorsulphonic acid
Kaava: C₁₀H₁₆O₄S
Kiehumispiste: 375 °C (1013 hPa)
Sulamispiste: 195°C
Tiheys: 1 g/cm³ (20 °C)
MDL numero: MFCD00064158
CAS-numero: 35963-20-3
EINECS: 252-817-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c3a7acebaf055f623aa47ae81f12fdfdd75d15f&vid=6fdf6c8cb09fc7a185b7e2b1b3c760bba8089a66&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -20.0 to -23.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR