β-Syklodekstriini ≥98.0% (by HPLC)

Toimittaja: TCI

C0900-500G C0900-25G C0900-100G
TCIAC0900-500GEA 93.5 EUR
TCIAC0900-500G TCIAC0900-25G TCIAC0900-100G
β-Syklodekstriini ≥98.0% (by HPLC)
β-Syklodekstriini
Kaava: C₄₂H₇₀O₃₅
MDL numero: MFCD00078139
CAS-numero: 7585-39-9
EINECS: 231-493-2
Merck Index: 14,02718
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f87dbb3e5ab95b46156c93ca5152f29a440336ee&vid=f87dbb3e5ab95b46156c93ca5152f29a440336ee&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +160 to +166 deg (C=1, H2O) (calcd.on anh.substance)
Water max. 14.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR