α-Syklodekstriini ≥98.0% (by HPLC)

Toimittaja: TCI

C0776-25G C0776-10G C0776-100G
TCIAC0776-25GEA 85.8 EUR
TCIAC0776-25G TCIAC0776-10G TCIAC0776-100G
α-Syklodekstriini ≥98.0% (by HPLC)
α-Syklodekstriini
Kaava: C₃₆H₆₀O₃₀
MDL numero: MFCD00078207
CAS-numero: 10016-20-3
EINECS: 233-007-4
Merck Index: 14,02718
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcd8a00adfd00cf016e76d85f11303fb8f62d544&vid=dcd8a00adfd00cf016e76d85f11303fb8f62d544&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +146.0 to +153.0 deg (C=1, H2O) (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR