Cholesterol hydrogen succinate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI

C0674-10G
TCIAC0674-10GEA 211 EUR
TCIAC0674-10G
Cholesterol hydrogen succinate ≥97.0% (by titrimetric analysis)
Cholesterol hydrogen succinate
Kaava: C₃₁H₅₀O₄
MDL numero: MFCD00037705
CAS-numero: 1510-21-0
EINECS: 216-148-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 175.0 to 181.0 deg-C
Specific rotation [a]20/D -32.0 to -36.0 deg (C=2, CHCl3)
Solubility in hot EtOH very faint turbidity

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR