Cholesterol Hydrocinnamate ≥95.0% (by GC)

Toimittaja: TCI

C0619-10G
TCIAC0619-10GEA 81.4 EUR
TCIAC0619-10G
Cholesterol Hydrocinnamate ≥95.0% (by GC)
Cholesterol Hydrocinnamate
Kaava: C₃₆H₅₄O₂
MDL numero: MFCD00003640
CAS-numero: 14914-99-9
EINECS: 238-983-5

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 108.0 to 112.0 deg-C
Specific rotation [a]20/D -17.0 to -20.0 deg (C=5, THF)
Solubility in THF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR