Phosphonitrilic chloride trimer ≥98.0% (by GC)

Toimittaja: TCI
Vaara

C0584-25G C0584-250G
TCIAC0584-25GEA 50.6 EUR
TCIAC0584-25G TCIAC0584-250G
Phosphonitrilic chloride trimer ≥98.0% (by GC)
Phosphonitrilic chloride trimer
Kaava: Cl₆N₃P₃
MDL numero: MFCD00006474
CAS-numero: 940-71-6
EINECS: 213-376-8
UN: 1759
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR