Crocein scarlet 3B

Toimittaja: TCI

C0531-25G
TCIAC0531-25GEA 18.7 EUR
TCIAC0531-25G
Crocein scarlet 3B
Crocein scarlet 3B
Kaava: C₂₂H₁₆N₄O₇S₂
MDL numero: MFCD00003907
CAS-numero: 5413-75-2
EINECS: 226-502-1

Order Now

Testitulosten erittely

Appearance Orange to Amber to Dark red powder to crystal
Lambda max. 508.0 to 512.0 nm (H2O)
Absorbance (E1%1cm) min. 490 (H2O, 508.0 to 512.0 nm)
Drying loss max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR