Crotonoyl chloride cis- and trans-mixture ≥95.0% (by GC)

Toimittaja: TCI
Vaara

C0424-25G C0424-500G C0424-100G
TCIAC0424-25GEA 35.2 EUR
TCIAC0424-25G TCIAC0424-500G TCIAC0424-100G
Crotonoyl chloride cis- and trans-mixture ≥95.0% (by GC)
Crotonoyl chloride
Kaava: C₄H₅ClO
Kiehumispiste: 125°C
Leimahduspiste: 35°C
MDL numero: MFCD00000734
CAS-numero: 10487-71-5
EINECS: 234-010-3
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8339966514f43cacc60735c7282228908eb24ad8&vid=d829646289c5a5ef5ee441ac38c1064fe11fd300&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR