Tricholine citrate ∼65% in water

Toimittaja: TCI

C0330-25G C0330-500G
TCIAC0330-25GEA 17.6 EUR
TCIAC0330-25G TCIAC0330-500G
Tricholine citrate ∼65% in water
Tricholine citrate
MDL numero: MFCD00059985
CAS-numero: 546-63-4
EINECS: 208-907-5

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Nonaqueous Titration) 60.0 to 66.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR