Choline bromide ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

C0328-25G C0328-500G
TCIAC0328-25GEA 56.1 EUR
TCIAC0328-25G TCIAC0328-500G
Choline bromide ≥98.0% (by titrimetric analysis)
Choline bromide
Kaava: C₅H₁₄BrNO
Sulamispiste: 298°C
MDL numero: MFCD00054244
CAS-numero: 1927-06-6
EINECS: 217-660-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7831c78332123fbdc59e945ae3777a6aa05f82ca&vid=7831c78332123fbdc59e945ae3777a6aa05f82ca&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR