Choline hydroxide 48 - 50% in water

Toimittaja: TCI
Vaara

C0326-25G C0326-500G C0326-100G
TCIAC0326-25GEA 17.6 EUR
TCIAC0326-25G TCIAC0326-500G TCIAC0326-100G
Choline hydroxide 48 - 50% in water
Choline hydroxide
Kaava: C₅H₁₅NO₂
Storage Temperature: Jääkaappi
MDL numero: MFCD00002831
CAS-numero: 123-41-1
EINECS: 204-625-1
UN: 3267
ADR: 8,II
Merck Index: 14,02204

Order Now

Testitulosten erittely

Appearance Colorless to Light red to Green clear liquid
Concentration (Neutralization titration) 46.5 to 50.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR