ε-Kaprolaktaami ≥99.0%, zone refined: number of passes 24

Toimittaja: TCI
Varoitus

C0022-1SAMPLE
TCIAC0022-1SAMPLEEA 388 EUR
TCIAC0022-1SAMPLE
ε-Kaprolaktaami ≥99.0%, zone refined: number of passes 24
ε-Kaprolaktaami

CAS Number: 105-60-2
MDL Number: MFCD00006936
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 70
Flash Point (°C): 130


Product Categories: Materials Chemistry, Highly Purified Reagents
Other Product Names / Synonyms: Cyclohexanoneisooxime, 2-Ketohexamethyleneimine, 2-Oxohexamethyleneimine

Kaava: C₆H₁₁NO
Kiehumispiste: 267 °C
Sulamispiste: 70 °C
Leimahduspiste: 130 °C
MDL numero: MFCD00006936
CAS-numero: 105-60-2
EINECS: 203-313-2

Order Now

Testitulosten erittely

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR