SB 225002 ≥98.0% (by HPLC)

Toimittaja: TCI

B6055-10MG B6055-50MG
TCIAB6055-10MGEA 90.2 EUR
TCIAB6055-10MG TCIAB6055-50MG
SB 225002 ≥98.0% (by HPLC)
SB 225002
Kaava: C₁₃H₁₀BrN₃O₄
MDL numero: MFCD00954637
CAS-numero: 182498-32-4

Order Now

Testitulosten erittely

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.30 to 12.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR