Belinostat ≥98.0% (by HPLC)

Toimittaja: TCI

Synonyms: (E)-N-Hydroxy-3-[3-(N-phenylsulphamoyl)phenyl]acrylamide, (E)-N-Hydroxy-3-(3-(N-phenylsulphamoyl)phenyl)acrylamide

B5888-100MG B5888-25MG
TCIAB5888-100MGEA 434 EUR
TCIAB5888-100MG TCIAB5888-25MG
Belinostat ≥98.0% (by HPLC)
Belinostat
Kaava: C₁₅H₁₄N₂O₄S
MDL numero: MFCD08064035
CAS-numero: 866323-14-0

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR