tert-Butoxytrimethylsilane ≥97.0% (by GC)

Toimittaja: TCI
Vaara

B5877-5ML B5877-25ML
TCIAB5877-5MLEA 55 EUR
TCIAB5877-5ML TCIAB5877-25ML
tert-Butoxytrimethylsilane ≥97.0% (by GC)
tert-Butoxytrimethylsilane
Kaava: C₇H₁₈OSi
Kiehumispiste: 102°C
MDL numero: MFCD00054893
CAS-numero: 13058-24-7
UN: 1993
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR