2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine ≥95.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

B5874-1ML B5874-5ML
TCIAB5874-1MLEA 28.6 EUR
TCIAB5874-1ML TCIAB5874-5ML
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine ≥95.0% (by GC, titration analysis)
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine
Kaava: C₈H₂₁NOSi
MDL numero: MFCD18206132
CAS-numero: 101711-55-1

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR