1,8-Bis(maleimido)-3,6-dioxaoctane ≥98.0% (by HPLC)

Toimittaja: TCI

B5667-250MG B5667-50MG
TCIAB5667-250MGEA 391 EUR
TCIAB5667-250MG TCIAB5667-50MG
1,8-Bis(maleimido)-3,6-dioxaoctane ≥98.0% (by HPLC)
1,8-Bis(maleimido)-3,6-dioxaoctane
Kaava: C₁₄H₁₆N₂O₆
Storage Temperature: Pakastin
MDL numero: MFCD01096749
CAS-numero: 115597-84-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.50 to 9.50 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR