Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Toimittaja: TCI
Varoitus

B5186-1G B5186-5G
TCIAB5186-1GEA 28.6 EUR
TCIAB5186-1G TCIAB5186-5G
Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Butylamine hydrobromide
Kaava: C₄H₁₁N·HBr
MDL numero: MFCD01874353
CAS-numero: 15567-09-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR