1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

B5002-1G B5002-5G
TCIAB5002-1GEA 72.6 EUR
TCIAB5002-1G TCIAB5002-5G
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
1-(1-Boc-piperidino)-1H-pyrazole-4-boronic acid pinacol ester
Kaava: C₁₉H₃₂BN₃O₄
Storage Temperature: Jääkaappi
MDL numero: MFCD11112131
CAS-numero: 877399-74-1

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR