3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine

Toimittaja: TCI

B4907-200MG B4907-1G
TCIAB4907-200MGEA 196 EUR
TCIAB4907-200MG TCIAB4907-1G
3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
3,3'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Kaava: C₃₆H₂₀N₂O₄
Sulamispiste: 340°C
CAS-numero: 1395881-58-9

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green powder to crystal
Elemental analysis (Nitrogen) 4.90 to 5.40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR