ω-Bromo-2,4-dihydroxyacetophenone ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

B4767-1G
TCIAB4767-1GEA 81.4 EUR
TCIAB4767-1G
ω-Bromo-2,4-dihydroxyacetophenone ≥97.0% (by titrimetric analysis)
ω-Bromo-2,4-dihydroxyacetophenone
Kaava: C₈H₇BrO₃
MDL numero: MFCD03789098
CAS-numero: 2491-39-6
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR