2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥99.0% (by HPLC, total nitrogen), purified by sublimation

Toimittaja: TCI

B4221-1G B4221-5G
TCIAB4221-1GEA 89.1 EUR
TCIAB4221-1G TCIAB4221-5G
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥99.0% (by HPLC, total nitrogen), purified by sublimation
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
Kaava: C₂₆H₂₆N₂O₂S
MDL numero: MFCD00005774
CAS-numero: 7128-64-5
EINECS: 230-426-4
Merck Index: 14,01014

Order Now

Testitulosten erittely

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR