6-Bromi-m-tolueenihappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

B4145-5G B4145-25G
TCIAB4145-5GEA 30.8 EUR
TCIAB4145-5G TCIAB4145-25G
6-Bromi-m-tolueenihappo ≥98.0% (by GC, titration analysis)
6-Bromi-m-tolueenihappo
Kaava: C₈H₇BrO₂
MDL numero: MFCD00079722
CAS-numero: 6967-82-4

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR