6-Bromi-o-tolueenihappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

B4097-1G B4097-5G
TCIAB4097-1GEA 53.9 EUR
TCIAB4097-1G TCIAB4097-5G
6-Bromi-o-tolueenihappo ≥98.0% (by GC, titration analysis)
6-Bromi-o-tolueenihappo
Kaava: C₈H₇BrO₂
MDL numero: MFCD01310788
CAS-numero: 90259-31-7

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR