tert-Butyl-N-allylcarbamate ≥98.0% (by GC)

Toimittaja: TCI

B3618-25G B3618-5G
TCIAB3618-25GEA 123 EUR
TCIAB3618-25G TCIAB3618-5G
tert-Butyl-N-allylcarbamate ≥98.0% (by GC)
tert-Butyl-N-allylcarbamate
Kaava: C₈H₁₅NO₂
MDL numero: MFCD00191870
CAS-numero: 78888-18-3

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR