[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

B3374-1G B3374-5G
TCIAB3374-1GEA 46.2 EUR
TCIAB3374-1G TCIAB3374-5G
[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride ≥98.0% (by titrimetric analysis)
[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride
Kaava: C₃₄H₂₈Cl₂CoFeP₂
MDL numero: MFCD03427161
CAS-numero: 67292-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ef9079ae14971c3ce06cd15d94dbc3155a3bbc5&vid=f24368b5da4d6ddae897499849dc0b5470e78825&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Yellow to Brown to Dark green powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR