1-Boc-piperidine-4-carboxylic acid ≥96.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

B3241-25G B3241-5G
TCIAB3241-25GEA 246 EUR
TCIAB3241-25G TCIAB3241-5G
1-Boc-piperidine-4-carboxylic acid ≥96.0% (by GC, titration analysis)
1-Boc-piperidine-4-carboxylic acid
Kaava: C₁₁H₁₉NO₄
MDL numero: MFCD00076999
CAS-numero: 84358-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7effe94b45b21cd7e51f524d3a4810580097f2e4&vid=7effe94b45b21cd7e51f524d3a4810580097f2e4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR