2-Bromi-5-joditolueeni ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

B3099-25G B3099-5G
TCIAB3099-25GEA 237 EUR
TCIAB3099-25G TCIAB3099-5G
2-Bromi-5-joditolueeni ≥98.0% (by GC)
2-Bromi-5-joditolueeni
Kaava: C₇H₆BrI
MDL numero: MFCD00070749
CAS-numero: 202865-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b05691f361fe11df27a9e67fdfcc2f67a2a6946&vid=6b05691f361fe11df27a9e67fdfcc2f67a2a6946&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR